Inositol, 98+%
Inositol, 98+%
Inositol, 98+%
Thermo Scientific Chemicals
Inositol, 98+%
CAS: 87-89-8 | C6H12O6 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA122265000
Price (MYR)
1,151.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,151.00
BOT
Add to cart
Specifications
Chemical Name or MaterialInositol
Name Note98+%
CAS87-89-8
Melting Point220°C to 228°C