Indole-5-carboxylic acid, 98%
Indole-5-carboxylic acid, 98%
Indole-5-carboxylic acid, 98%
Indole-5-carboxylic acid, 98%
Thermo Scientific Chemicals
Indole-5-carboxylic acid, 98%
CAS: 1670-81-1 | C9H6NO2 | 160.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA122210050
Price (MYR)
1,136.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,136.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIndole-5-carboxylic acid
Name Note98%
CAS1670-81-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: