Indole-3-carbinol, 97%
Indole-3-carbinol, 97%
Indole-3-carbinol, 97%
Indole-3-carbinol, 97%
Thermo Scientific Chemicals
Indole-3-carbinol, 97%
CAS: 700-06-1 | C9H9NO | 147.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA122190250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIndole-3-carbinol
Name Note99%
CAS700-06-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement