Indole, 99+%
Indole, 99+%
Indole, 99+%
Thermo Scientific Chemicals
Indole, 99+%
CAS: 120-72-9 | C8H7N | 117.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
View all
Glass Bottle
Catalog number FSA122151000
Price (MYR)
582.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
582.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIndole
Name Note98+%
CAS120-72-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.