3-Hydroxytyramine Hydrochloride, 99%
3-Hydroxytyramine Hydrochloride, 99%
3-Hydroxytyramine Hydrochloride, 99%
3-Hydroxytyramine Hydrochloride, 99%
Thermo Scientific Chemicals
3-Hydroxytyramine Hydrochloride, 99%
CAS: 62-31-7 | C8H12ClNO2 | 189.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA122001000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Hydroxytyramine hydrochloride
CAS62-31-7
CAS Min %98.5
CAS Max %100.0
Melting Point241°C to 243°C