3-Hydroxytetrahydrofuran, 97%
3-Hydroxytetrahydrofuran, 97%
3-Hydroxytetrahydrofuran, 97%
3-Hydroxytetrahydrofuran, 97%
Thermo Scientific Chemicals
3-Hydroxytetrahydrofuran, 97%
CAS: 453-20-3 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA121940250
Price (MYR)
791.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
791.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Hydroxytetrahydrofuran
CAS453-20-3
CAS Min %96.0
ColorColorless to Yellow
Health Hazard 1GHS Signal Word: Warning