8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA121905000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material8-Hydroxyquinoline
Name Note99%
CAS148-24-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
May damage fertility or the unborn child.
Very toxic to aquatic life with long lasting effects.
View more
Indicator
RUO – Research Use Only