4-Hydroxyphenylacetic acid, 98%
4-Hydroxyphenylacetic acid, 98%
4-Hydroxyphenylacetic acid, 98%
4-Hydroxyphenylacetic acid, 98%
Thermo Scientific Chemicals
4-Hydroxyphenylacetic acid, 98%
CAS: 156-38-7 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA121710250
Price (MYR)
318.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
318.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Hydroxyphenylacetic acid
Name Note98%
CAS156-38-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning