5-Hydroxy-p-naphthoquinone, 97%
5-Hydroxy-p-naphthoquinone, 97%
5-Hydroxy-p-naphthoquinone, 97%
5-Hydroxy-p-naphthoquinone, 97%
Thermo Scientific Chemicals
5-Hydroxy-p-naphthoquinone, 97%
CAS: 481-39-0 | C10H6O3 | 174.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA121640050
Price (MYR)
1,154.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,154.00
BOT
Add to cart
Specifications
Chemical Name or Material5-Hydroxy-p-naphthoquinone
Name Note97%
CAS481-39-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger