2-Hydroxy-p-naphthoquinone, 99%
2-Hydroxy-p-naphthoquinone, 99%
2-Hydroxy-p-naphthoquinone, 99%
2-Hydroxy-p-naphthoquinone, 99%
Thermo Scientific Chemicals
2-Hydroxy-p-naphthoquinone, 99%
CAS: 83-72-7 | C10H6O3 | 174.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA121631000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Hydroxy-p-naphthoquinone
Name Note99%
CAS83-72-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning