2-Hydroxy-1-naphthaldehyde, tech.
2-Hydroxy-1-naphthaldehyde, tech.
2-Hydroxy-1-naphthaldehyde, tech.
2-Hydroxy-1-naphthaldehyde, tech.
Thermo Scientific Chemicals
2-Hydroxy-1-naphthaldehyde, tech.
CAS: 708-06-5 | C11H8O2 | 172.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA121605000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material2-Hydroxy-1-naphthaldehyde
Name Notetech.
CAS708-06-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Very toxic to aquatic life with long lasting effects.