3-Hydroxy-2-methyl-4-pyrone, 99%
3-Hydroxy-2-methyl-4-pyrone, 99%
3-Hydroxy-2-methyl-4-pyrone, 99%
3-Hydroxy-2-methyl-4-pyrone, 99%
Thermo Scientific Chemicals
3-Hydroxy-2-methyl-4-pyrone, 99%
CAS: 118-71-8 | C6H6O3 | 126.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA121551000
Price (MYR)
703.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
703.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Hydroxy-2-methyl-4-pyrone
CAS118-71-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0