5-(Hydroxymethyl)furfural, 98%
5-(Hydroxymethyl)furfural, 98%
5-(Hydroxymethyl)furfural, 98%
5-(Hydroxymethyl)furfural, 98%
Thermo Scientific Chemicals
5-(Hydroxymethyl)furfural, 98%
CAS: 67-47-0 | C6H6O3 | 126.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA121460250
Price (MYR)
3,856.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,856.00
EA
Add to cart
Specifications
Chemical Name or Material5-(Hydroxymethyl)furfural
CAS67-47-0
Health Hazard 1GHS Signal Word: Warning
Melting Point30.0°C to 34.0°C
ColorYellow