2-Hydroxy-4-methoxybenzophenone, 98%
2-Hydroxy-4-methoxybenzophenone, 98%
2-Hydroxy-4-methoxybenzophenone, 98%
2-Hydroxy-4-methoxybenzophenone, 98%
Thermo Scientific Chemicals
2-Hydroxy-4-methoxybenzophenone, 98%
CAS: 131-57-7 | C14H12O3 | 228.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA121361000
Price (MYR)
554.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
554.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Hydroxy-4-methoxybenzophenone
Name Note98%
CAS131-57-7
CAS Min %97.5
CAS Max %100.0