2'-Hydroxy-4'-methoxyacetophenone, 99%
2'-Hydroxy-4'-methoxyacetophenone, 99%
2'-Hydroxy-4'-methoxyacetophenone, 99%
2'-Hydroxy-4'-methoxyacetophenone, 99%
Thermo Scientific Chemicals
2'-Hydroxy-4'-methoxyacetophenone, 99%
CAS: 552-41-0 | C9H10O3 | 166.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA121330100
Price (MYR)
467.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
467.00
EA
Add to cart
Specifications
Chemical Name or Material2'-Hydroxy-4'-methoxyacetophenone
CAS552-41-0
CAS Min %98.5
CAS Max %100.0
Health Hazard 2GHS H Statement: