5-Hydroxyindole, 97%
5-Hydroxyindole, 97%
5-Hydroxyindole, 97%
5-Hydroxyindole, 97%
Thermo Scientific Chemicals
5-Hydroxyindole, 97%
CAS: 1953-54-4 | C8H7NO | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA121270010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material5-Hydroxyindole
CAS1953-54-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0