N-(2-Hydroxyethyl)piperazine, 98.5%
N-(2-Hydroxyethyl)piperazine, 98.5%
N-(2-Hydroxyethyl)piperazine, 98.5%
N-(2-Hydroxyethyl)piperazine, 98.5%
Thermo Scientific Chemicals
N-(2-Hydroxyethyl)piperazine, 98.5%
CAS: 103-76-4 | C6H14N2O | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA121191000
Price (MYR)
322.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
322.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN-(2-Hydroxyethyl)piperazine
Name Note98.5%
CAS103-76-4
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger