N-(2-Hydroxyethyl)morpholine, 99%
N-(2-Hydroxyethyl)morpholine, 99%
N-(2-Hydroxyethyl)morpholine, 99%
N-(2-Hydroxyethyl)morpholine, 99%
Thermo Scientific Chemicals
N-(2-Hydroxyethyl)morpholine, 99%
CAS: 622-40-2 | C6H13NO2 | 131.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA121181000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN-(2-Hydroxyethyl)morpholine
Name Note99%
CAS622-40-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning