7-Hydroxycoumarin, 99%
7-Hydroxycoumarin, 99%
7-Hydroxycoumarin, 99%
7-Hydroxycoumarin, 99%
Thermo Scientific Chemicals
7-Hydroxycoumarin, 99%
CAS: 93-35-6 | C9H6O3 | 162.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA121110250
Price (MYR)
466.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
466.00
BOT
Add to cart
Specifications
Chemical Name or Material7-Hydroxycoumarin
Name Note99%
CAS93-35-6
CAS Min %98.5
CAS Max %100.0