4-Hydroxybenzoic Acid Hydrazide, 98%
4-Hydroxybenzoic Acid Hydrazide, 98%
4-Hydroxybenzoic Acid Hydrazide, 98%
4-Hydroxybenzoic Acid Hydrazide, 98%
Thermo Scientific Chemicals
4-Hydroxybenzoic Acid Hydrazide, 98%
CAS: 5351-23-5 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number FSA121005000
Price (MYR)
4,144.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
4,144.00
EA
Add to cart
Specifications
Chemical Name or Material4-Hydroxybenzoic acid hydrazide
CAS5351-23-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0