2-Hydroxybenzimidazole, 97%
2-Hydroxybenzimidazole, 97%
2-Hydroxybenzimidazole, 97%
2-Hydroxybenzimidazole, 97%
Thermo Scientific Chemicals
2-Hydroxybenzimidazole, 97%
CAS: 615-16-7 | C7H6N2O | 134.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA120970250
Price (MYR)
746.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
746.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Hydroxybenzimidazole
Name Note97%
CAS615-16-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning