3-Hydroxybenzaldehyde, 98.5%
3-Hydroxybenzaldehyde, 98.5%
3-Hydroxybenzaldehyde, 98.5%
3-Hydroxybenzaldehyde, 98.5%
Thermo Scientific Chemicals
3-Hydroxybenzaldehyde, 98.5%
CAS: 100-83-4 | C7H6O2 | 122.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA120965000
Price (MYR)
1,368.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,368.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Hydroxybenzaldehyde
Name Note98.5%
CAS100-83-4
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning