2-Hydrazinopyridine, 98%
2-Hydrazinopyridine, 98%
2-Hydrazinopyridine, 98%
2-Hydrazinopyridine, 98%
Thermo Scientific Chemicals
2-Hydrazinopyridine, 98%
CAS: 4930-98-7 | C5H7N3 | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA120870050
Price (MYR)
665.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
665.00
EA
Add to cart
Specifications
Chemical Name or Material2-Hydrazinopyridine
CAS4930-98-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0