Humic acid, sodium salt, 45-70%, technical, (as humic acid)
Humic acid, sodium salt, 45-70%, technical, (as humic acid)
Humic acid, sodium salt, 45-70%, technical, (as humic acid)
Humic acid, sodium salt, 45-70%, technical, (as humic acid)
Thermo Scientific Chemicals
Humic acid, sodium salt, 45-70%, technical, (as humic acid)
CAS: 68131-04-4 | C9H8Na2O4 | 226.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA120861000
Price (MYR)
313.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
313.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHumic acid, sodium salt
Name Notetech., 50-60% (as humic acid)
CAS68131-04-4
Melting Point>300.0°C
ColorBlack