Hexyl alcohol, 98%, pure
Hexyl alcohol, 98%, pure
Hexyl alcohol, 98%, pure
Hexyl alcohol, 98%, pure
Hexyl alcohol, 98%, pure
CAS: 111-27-3 | C6H14O | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA120791000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHexyl alcohol
Acidity0.02mg KOH/g max.
CAS111-27-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Flammable liquid and vapor.