1-Hexene, 99%, AcroSeal™
1-Hexene, 99%, AcroSeal™
1-Hexene, 99%, AcroSeal™
1-Hexene, 99%, AcroSeal™
1-Hexene, 99%, AcroSeal™
Thermo Scientific Chemicals
1-Hexene, 99%, AcroSeal™
CAS: 592-41-6 | C6H12 | 84.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA120751000
Price (MYR)
426.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
426.00
EA
Add to cart
Specifications
Chemical Name or Material1-Hexene
CAS592-41-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0