Hexanoic acid, 99%
Hexanoic acid, 99%
Hexanoic acid, 99%
Thermo Scientific Chemicals
Hexanoic acid, 99%
CAS: 142-62-1 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
250 g
2.5 kg
Packaging:
Glass bottle
Catalog number FSA120705000
Price (MYR)
413.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
413.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHexanoic acid
Acidity473 to 493mg KOH/g
CAS142-62-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement