1,6-Hexanediamine, 99.5+%
1,6-Hexanediamine, 99.5+%
1,6-Hexanediamine, 99.5+%
1,6-Hexanediamine, 99.5+%
Thermo Scientific Chemicals
1,6-Hexanediamine, 99.5+%
CAS: 124-09-4 | C6H16N2 | 116.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA120645000
Price (MYR)
508.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
508.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 6-Hexanediamine
CAS124-09-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with wate