2,4-Hexadienoic acid, 99%
2,4-Hexadienoic acid, 99%
2,4-Hexadienoic acid, 99%
2,4-Hexadienoic acid, 99%
Thermo Scientific Chemicals
2,4-Hexadienoic acid, 99%
CAS: 110-44-1 | C6H8O2 | 112.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA120535000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 4-Hexadienoic acid
Name Note99%
CAS110-44-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning