1-Hexadecylamine, 90%
1-Hexadecylamine, 90%
1-Hexadecylamine, 90%
1-Hexadecylamine, 90%
Thermo Scientific Chemicals
1-Hexadecylamine, 90%
CAS: 143-27-1 | C16H35N | 241.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA120515000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Hexadecylamine
CAS143-27-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for