1-Hexadecanol, 96%
1-Hexadecanol, 96%
1-Hexadecanol, 96%
Thermo Scientific Chemicals
1-Hexadecanol, 96%
CAS: 36653-82-4 | C16H34O | 242.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
1 kg
2.5 kg
Catalog number FSA120480100
Price (MYR)
2,566.00
EA
Add to cart
Quantity:
10 kg
Request bulk or custom format
Price (MYR)
2,566.00
EA
Add to cart
Specifications
Chemical Name or Material1-Hexadecanol
CAS36653-82-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Health Hazard 3GHS P Statement