n-Hexadecane, 99%, pure
n-Hexadecane, 99%, pure
n-Hexadecane, 99%, pure
n-Hexadecane, 99%, pure
Thermo Scientific Chemicals
n-Hexadecane, 99%, pure
n-Hexadecane, 99%, C16H34, CAS Number-544-76-3, n-cetane, n-hexadecane, hexadecan, hexadekan, cetan, cetane, ccris 5833, zetan, pentadecane, methyl, unii-f8z00shp6q, 2.5L, 98.5% min. (GC), 01,172, 287.0 deg.C, 100.0, CHEBI:45296, Colorless, 0.7730g/mL, 208-878-9, 135 deg.C, 226.44, Pure | CAS: 544-76-3 | C16H34 | 226.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Catalog number FSA120465000
Price (MYR)
1,101.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,101.00
BOT
Add to cart
Specifications
Chemical Name or Materialn-Hexadecane
CAS544-76-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0