3-Heptanone, 98%
3-Heptanone, 98%
3-Heptanone, 98%
Thermo Scientific Chemicals
3-Heptanone, 98%
CAS: 106-35-4 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA120382500
Price (MYR)
502.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
502.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Heptanone
CAS106-35-4
Health Hazard 1GHS Signal Word: Warning
Melting Point-39.0°C
ColorYellow