2-Heptanol, 99+%
2-Heptanol, 99+%
2-Heptanol, 99+%
Thermo Scientific Chemicals
2-Heptanol, 99+%
CAS: 543-49-7 | C7H16O | 116.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
Packaging:
Glass bottle
Catalog number FSA120372500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Heptanol
Name Note99+%
CAS543-49-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning