n-Heptane, 99+%, pure
n-Heptane, 99+%, pure
n-Heptane, 99+%, pure
n-Heptane, 99+%, pure
Thermo Scientific Chemicals
n-Heptane, 99+%, pure
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
Catalog number FSA120340050
Price (MYR)
1,248.00
LT
Add to cart
Quantity:
5 L
Request bulk or custom format
Price (MYR)
1,248.00
LT
Add to cart
Specifications
Chemical Name or Materialn-Heptane
Melting Point-91.0°C
CAS142-82-5
CAS Min %99.0