Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2)
Guanidine hydrochloride, 98%, with &lt;0.4% anticaking agent (SiO2)
Guanidine hydrochloride, 98%, with &lt;0.4% anticaking agent (SiO2)
Guanidine hydrochloride, 98%, with &lt;0.4% anticaking agent (SiO2)
Thermo Scientific Chemicals
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2)
Guanidine hydrochloride, 98%, with <0.4% anticaking agent (SiO2), Quantity: 1kg, Packaging: Plastic bottle, White to Yellow, Melting Point: 179.0 deg.C to 186.0 deg.C, Molecular Weight: 95.53, Percent Purity: 98%, Assay Percent Range: 0.5% max. Ammonium chloride (Argentometry) | CAS: 50-01-1 | CH5N3·HCl | 95.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA120232500
Price (MYR)
262.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
262.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGuanidine hydrochloride
CAS50-01-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Health Hazard 3GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse