Guanidine Carbonate, 99+%
Guanidine Carbonate, 99+%
Guanidine Carbonate, 99+%
Guanidine Carbonate, 99+%
Thermo Scientific Chemicals
Guanidine Carbonate, 99+%
CAS: 593-85-1 | 0·5 H2CO3 | 90.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number FSA120222500
Price (MYR)
281.00
BOT
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
281.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGuanidine carbonate
Name Note99+%
CAS593-85-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: