Guaiacol, 99+%
Guaiacol, 99+%
Guaiacol, 99+%
Thermo Scientific Chemicals
Guaiacol, 99+%
CAS: 90-05-1 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA120192500
Price (MYR)
344.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
344.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGuaiacol
CAS90-05-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con