Glycine anhydride, 98%
Glycine anhydride, 98%
Glycine anhydride, 98%
Glycine anhydride, 98%
Thermo Scientific Chemicals
Glycine anhydride, 98%
CAS: 106-57-0 | C4H6N2O2 | 114.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA120081000
Price (MYR)
1,413.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,413.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGlycine anhydride
Name Note98%
CAS106-57-0
Melting Point>300.0°C
ColorWhite to Yellow