Glutathione, 98%, for analysis, reduced
Glutathione, 98%, for analysis, reduced
Glutathione, 98%, for analysis, reduced
Glutathione, 98%, for analysis, reduced
Thermo Scientific Chemicals
Glutathione, 98%, for analysis, reduced
CAS: 70-18-8 | C10H17N3O6S | 307.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 g
5 g
25 g
Catalog number FSA120001000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGlutathione
Name NoteFor analysis, Reduced
CAS70-18-8
Melting Point182.0°C to 192.0°C