Glutaric anhydride, 95%
Glutaric anhydride, 95%
Glutaric anhydride, 95%
Glutaric anhydride, 95%
Thermo Scientific Chemicals
Glutaric anhydride, 95%
CAS: 108-55-4 | C5H6O3 | 114.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA119971000
Price (MYR)
522.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
522.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGlutaric anhydride
Name Note97%
CAS108-55-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement