L(+)-Glutamine, 99%
L(+)-Glutamine, 99%
L(+)-Glutamine, 99%
Thermo Scientific Chemicals
L(+)-Glutamine, 99%
CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number FSA119955000
Price (MYR)
1,668.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,668.00
BOT
Add to cart
Specifications
Chemical Name or MaterialL(+)-Glutamine
Melting Point185.0°C
Name Note99%
CAS56-85-9
Health Hazard 1LIGHT SENSITIVE