L(+)-Glutamic acid monosodium salt monohydrate, 99%
L(+)-Glutamic acid monosodium salt monohydrate, 99%
L(+)-Glutamic acid monosodium salt monohydrate, 99%
L(+)-Glutamic acid monosodium salt monohydrate, 99%
Thermo Scientific Chemicals
L(+)-Glutamic acid monosodium salt monohydrate, 99%
CAS: 6106-04-03 | C5H8NNaO4·H2O | 187.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic Bottle
Catalog number FSA119940010
Price (MYR)
399.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
399.00
BOT
Add to cart
Specifications
Chemical Name or MaterialL(+)-Glutamic acid monosodium salt monohydrate
Melting Point232.0°C
CAS6106-04-03
ColorWhite
Chloride2000ppm max