Girard's Reagent T, 99%
Girard's Reagent T, 99%
Girard's Reagent T, 99%
Girard's Reagent T, 99%
Thermo Scientific Chemicals
Girard's Reagent T, 99%
CAS: 123-46-6 | C5H14ClN3O | 167.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number FSA119872500
Price (MYR)
914.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
914.00
EA
Add to cart
Specifications
Chemical Name or MaterialGirard's Reagent T
CAS123-46-6
Health Hazard 1Warning
Health Hazard 2GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.