Gibberellic Acid, 90%
Gibberellic Acid, 90%
Gibberellic Acid, 90%
Gibberellic Acid, 90%
Thermo Scientific Chemicals
Gibberellic Acid, 90%
CAS: 77-06-5 | C19H22O6 | 346.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA119860250
Price (MYR)
3,351.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,351.00
EA
Add to cart
Specifications
Chemical Name or MaterialGibberellic acid
CAS77-06-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.