Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol
Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol
Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol
Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol
Thermo Scientific Chemicals
Formaldehyde, 37 wt% sol. in water, stab. with 5-15% methanol
Formaldehyde, formalin, CAS # 50-00-0, is a colorless volatile used as a histologic fixative and disinfectant. | CAS: 50-00-0 | CH2O | 30.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Plastic bottle
Catalog number FSA119690025
Price (MYR)
429.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
429.00
EA
Add to cart
Chemical Identifiers
CAS50-00-0, 7732-18-5, 67-56-1
Specifications
Chemical Name or MaterialFormaldehyde
Name Note37 wt% Solution in Water, Stabilized with 10 to 15% Methanol
CAS50-00-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes damage to organs.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause cancer.
Fl
View more
General description
  • Formaldehyde (formalin) is an organic compound used for crosslinking proteins in biological samples such as tissue sections, culture cells, and whole specimens
  • Formaldehyde is an intermediate in the combustion of methane and can exist in many forms
  • It is a volatile, colorless liquid; that can be stored long-term in appropriate containers
  • Comes stabilized in solution with methanol
Applications
  • It May be used in pain research for induction of pain and inflammation
  • Monomeric formaldehyde is formulated especially for laboratory applications
  • Commonly used in RNA isolation as a denaturant
RUO – Research Use Only