4-Fluorophenylhydrazine hydrochloride, 97%
4-Fluorophenylhydrazine hydrochloride, 97%
4-Fluorophenylhydrazine hydrochloride, 97%
4-Fluorophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals
4-Fluorophenylhydrazine hydrochloride, 97%
CAS: 823-85-8 | C6H8ClFN2 | 162.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number FSA119590250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Fluorophenylhydrazine hydrochloride
CAS823-85-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0