1-Fluoro-4-nitrobenzene, 99%
1-Fluoro-4-nitrobenzene, 99%
1-Fluoro-4-nitrobenzene, 99%
1-Fluoro-4-nitrobenzene, 99%
Thermo Scientific Chemicals
1-Fluoro-4-nitrobenzene, 99%
CAS: 350-46-9 | C6H4FNO2 | 141.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA119491000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Fluoro-4-nitrobenzene
Name Note99%
CAS350-46-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning