1-Fluoro-2-nitrobenzene, 99%
1-Fluoro-2-nitrobenzene, 99%
1-Fluoro-2-nitrobenzene, 99%
1-Fluoro-2-nitrobenzene, 99%
Thermo Scientific Chemicals
1-Fluoro-2-nitrobenzene, 99%
CAS: 1493-27-2 | C6H4FNO2 | 141.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Packaging:
Glass bottle
Catalog number FSA119480100
Price (MYR)
236.00
BOT
Add to cart
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
236.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Fluoro-2-nitrobenzene
CAS1493-27-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0