3-Fluorobenzoic acid, 99%
3-Fluorobenzoic acid, 99%
3-Fluorobenzoic acid, 99%
3-Fluorobenzoic acid, 99%
Thermo Scientific Chemicals
3-Fluorobenzoic acid, 99%
CAS: 455-38-9 | C7H5FO2 | 140.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA119370250
Price (MYR)
402.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
402.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Fluorobenzoic acid
Name Note99%
CAS455-38-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning